Learn More
Microsolv Technology Corporation Rsa Vial Crimp 100/Pk
Catalog No. NC0481883
Encompass_Preferred
Vials, Snap Top, Glass. Clear, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK
Catalog No. NC0481883 Supplier Microsolv Technology Corporation Supplier No. 9509CWCVRS
Vials, Snap Top, Glass. Clear, 2ml, with Write-On Patch and fill lines. An 11mm snap-ring and 12x32mm outer dimensions. For use as an autosampler vial. RSA Brand. 100/PK