Learn More
Aviva Systems Biology Corp RSAD1 Antibody - middle region (ARP57212_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57212_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RSAD1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the middle region of human RSAD1. Alternate names: FLJ11164, FLJ20975ApplicationWB. Species reactivity: Cow, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-286-281