Learn More
New England Biolabs, Inc. RsaI – 1000 units
SDP
Catalog No. 50812073
Encompass
A restriction endonuclease that recognizes the sequence GT_^AC.
Catalog No. 50-812-073 Supplier New England Biolabs, Inc. Supplier No. R0167S
A restriction endonuclease that recognizes the sequence GT_^AC.