Learn More
Aviva Systems Biology Corp RSBN1 Antibody - N-terminal region (ARP57222_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP57222_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RSBN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human RSBN1. Alternate names: DKFZp781E21150, ROSBIN, RP11-324J2.1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-53875