Learn More
STEMCELL Technologies RSC-133, Size: 10 mg
SDP
Supplier:  STEMCELL Technologies 73392
Encompass
Reprogramming stimulating compound (RSC)-133 is an indole derivative that specifically inhibits DNA methyltransferase DNMT1 (Lee et al.).REPROGRAMMING· Increases kinetics and efficiency of reprogramming human foreskin fibroblasts to induced pluripotent stem cells, when used with reprogramming factors OCT4, SOX2, KLF4, and c-MYC (Lee et al.). MAINTENANCE & SELF-RENEWAL· Supports the maintenance of human pluripotent stem cells in unconditioned medium (Lee et al.).
Catalog No. 50-197-6070