Learn More
Enzo Life Sciences rSec6 monoclonal antibody (9H5) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMSV021D
Encompass_Preferred
Clone: 9H5. Host: Mouse. Isotype: IgG2a. Immunogen: Recombinant rat Sec6. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Dog, Chicken, Bovine. Applications: ICC, IHC (PS), WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7699