Learn More
Enzo Life Sciences rSec8 monoclonal antibody (14G1) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMSV016D
Encompass_Preferred
Clone: 14G1. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat rSec8. Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Sheep, Rabbit, Porcine, Monkey, Hamster, Guinea pig, Fish, Dog, Chicken, Bovine. Applications: ICC, IHC (PS), IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7696