Learn More
Aviva Systems Biology Corp RSF1 Antibody - middle region (ARP48794_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP48794_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against RSF1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human RSF1. Alternate names: HBXAP, RSF-1, XAP8, p325ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59925