Learn More
NABERTHERM INC These tube furnaces can be used for horizontal operation. The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes.
Catalog No. NC2332311
Encompass_Preferred
These tube furnaces can be used for horizontal operation. The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes (e.g. working tubes made of different materials).Using a wide range of accessories, these professional tube furnaces can be optimally tailored for your process. By adding different gas supply packages, you can work in a protective gas atmosphere, with gases or in a vacuum. In addition to the convenient standard controllers, modern PLC controls can also be used to control the process.
Catalog No. NC2332311 Supplier NABERTHERM INC Supplier No. RS322L3AN480
Add to Cart
Add to Cart
Request a Quote
These tube furnaces can be used for horizontal operation. The split-type design makes it easy to change the working tube. It allows for a comfortable exchange of various working tubes (e.g. working tubes made of different materials).Using a wide range of accessories, these professional tube furnaces can be optimally tailored for your process. By adding different gas supply packages, you can work in a protective gas atmosphere, with gases or in a vacuum. In addition to the convenient standard controllers, modern PLC controls can also be used to control the process.