Learn More
Carnabio Usa Inc RSK1(RPS6KA1)/10UG/FULL-LENGTH
Supplier:  Carnabio Usa Inc 0114910
Encompass
RSK1(RPS6KA1), Full-length, Wild type, Amino Acid 1-735, NP_002944.2, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0978