Learn More
Sino Biological Recombinant Human Ros1 Protein (F2004C, GST Tag), Active
Catalog No. 502573258
Encompass_Preferred
Recombinant human ROS1 (F2004C) (1881-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.
Catalog No. 50-257-3258 Supplier Sino Biological Supplier No. R1412FG500
Recombinant human ROS1 (F2004C) (1881-end) was expressed by baculovirus in Sf9 insect cells using an N-terminal GST tag.