Learn More
Eurofins Discoverx Rsk3, active, 10 µg. (14-462)
Supplier:  Eurofins Discoverx 14462
Encompass
N-terminal 6xHis-tagged recombinant, human full length RSK3. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8827