Learn More
Carnabio Usa Inc RSK4(RPS6KA6)/5UG/FULL-LENGTH
Catalog No. 507540619
Encompass
RSK4(RPS6KA6), Full-length, Wild type, Amino Acid 1-745, NP_055311.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0619 Supplier Carnabio Usa Inc Supplier No. 011525
RSK4(RPS6KA6), Full-length, Wild type, Amino Acid 1-745, NP_055311.1, Expressed in Insect (sf21); 5microg