Learn More
Abcam RSL3, 5MG
Supplier:  Abcam AB2854375MG
Encompass_Preferred
MW 440.9 Da, Purity >=98%. RSL3 is a glutathione peroxidase 4 (GPX4) inhibitor and an inducer of ferroptosis. RSL3 increases lipid peroxidase levels in vitro and inhibits tumor formation and progression in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-220-5535