Learn More
Selleck Chemical LLC RSL3
SDP
Catalog No. 5000000067
Encompass_Preferred
RSL3 ((1S 3R)-RSL3) is a ferroptosis activator in a VDAC-independent manner exhibiting selectivity for tumor cells bearing oncogenic RAS RSL3 binds inactivates GPX4 and thus mediates GPX4-regulated ferroptosis
Catalog No. 50-000-00067 Supplier Selleck Chemical LLC Supplier No. S81551G
RSL3 ((1S 3R)-RSL3) is a ferroptosis activator in a VDAC-independent manner exhibiting selectivity for tumor cells bearing oncogenic RAS RSL3 binds inactivates GPX4 and thus mediates GPX4-regulated ferroptosis