Learn More
Medchemexpress LLC HY-100218A 2mg Medchemexpress, RSL3 CAS:1219810-16-8 Purity:>98%
SDP
Catalog No. 502028611
Encompass_Preferred
Medchemexpress, HY-100218A 2mg RSL3 CAS:1219810-16-8 RSL3 ((1S,3R)-RSL3) is an inhibitor of glutathione peroxidase 4 (GPX4) (ferroptosis activator), reduces the expression of GPX4 protein, and induces ferroptotic death of head and neck cancer cell. RSL3 increases the expression of p62 and Nrf2 and inactivates Keap1 in HN3-rslR cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8611 Supplier Medchemexpress LLC Supplier No. HY100218A2MG
Medchemexpress, HY-100218A 2mg RSL3 CAS:1219810-16-8 RSL3 ((1S,3R)-RSL3) is an inhibitor of glutathione peroxidase 4 (GPX4) (ferroptosis activator), reduces the expression of GPX4 protein, and induces ferroptotic death of head and neck cancer cell. RSL3 increases the expression of p62 and Nrf2 and inactivates Keap1 in HN3-rslR cells. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.