Learn More
Cayman Chemical (R)-SLV 319, Purity: ≥98%, CAS: 656827-86-0, 5 mg
SDP
Catalog No. 502890914
Encompass_Preferred
(R)-SLV 319 is the inactive enantiomer of SLV 319 with 100-fold less affinity for the CB1 receptor than (S)-SLV 319
Catalog No. 50-289-0914 Supplier Cayman Chemical Supplier No. 100092275 MG
(R)-SLV 319 is the inactive enantiomer of SLV 319 with 100-fold less affinity for the CB1 receptor than (S)-SLV 319