Learn More
Cayman Chemical RSLV 319 Neurochemicals
SDP
Supplier:  Cayman Chemical 100092271 MG
Encompass_Preferred
(R)-SLV 319 is the inactive enantiomer of SLV 319 with 100-fold less affinity for the CB1 receptor than (S)-SLV 319
Catalog No. 50-288-7858