Learn More
Medchemexpress LLC Rsm-932a (TCD-717) | 850807-63-5 | 98.4% | 877.53 g/mol | C46H38Br2Cl2N4 | 25 MG
SDP
Catalog No. 5000211356
Encompass_Preferred
RSM-932A (TCD-717) is a small-molecule choline kinase alpha (ChoKα) inhibitor provided for research use. Supplied as a solid with high reported purity, it is intended for in vitro biochemical and cellular studies; follow the manufacturer's safety and storage guidance.
Catalog No. 50-002-11356 Supplier Medchemexpress LLC Supplier No. HY10114425MG
RSM-932A (TCD-717) is a small-molecule choline kinase alpha (ChoKα) inhibitor provided for research use. Supplied as a solid with high reported purity, it is intended for in vitro biochemical and cellular studies; follow the manufacturer's safety and storage guidance.