Learn More
Medchemexpress LLC Rsm3 Tfa | 98.8% | 3049.66 | 1 MG
SDP
Catalog No. 5000425115
Encompass_Preferred
This product is a stapled peptide that acts as an inhibitor of METTL3-METTL14, with a Kd of 3.10 μM. It has been shown to inhibit tumor growth and induce cell apoptosis, making it useful for cancer research.
  • It is a stapled peptide.
  • It is a METTL3-METTL14 inhibitor.
  • It inhibits tumor growth.
  • It induces cell apoptosis.
  • It is suitable for cancer research.
Catalog No. 50-004-25115 Supplier Medchemexpress LLC Supplier No. HYP10387A1MG
This product is a stapled peptide that acts as an inhibitor of METTL3-METTL14, with a Kd of 3.10 μM. It has been shown to inhibit tumor growth and induce cell apoptosis, making it useful for cancer research.
  • It is a stapled peptide.
  • It is a METTL3-METTL14 inhibitor.
  • It inhibits tumor growth.
  • It induces cell apoptosis.
  • It is suitable for cancer research.