Learn More
Medchemexpress LLC RSM3 TFA | 98.83% | 3049.66 | 5 MG
SDP
Catalog No. 5000425396
Encompass_Preferred
RSM3 TFA, a stapled peptide, is a METTL3-METTL14 inhibitor with a Kd of 3.10 μM. It inhibits tumor growth and induces cell apoptosis. RSM3 TFA can be used for the study of cancer.
  • Inhibits tumor growth
  • Induces cell apoptosis
  • Used for the study of cancer
  • Stapled peptide
  • METTL3-METTL14 inhibitor
Catalog No. 50-004-25396 Supplier Medchemexpress LLC Supplier No. HYP10387A5MG
RSM3 TFA, a stapled peptide, is a METTL3-METTL14 inhibitor with a Kd of 3.10 μM. It inhibits tumor growth and induces cell apoptosis. RSM3 TFA can be used for the study of cancer.
  • Inhibits tumor growth
  • Induces cell apoptosis
  • Used for the study of cancer
  • Stapled peptide
  • METTL3-METTL14 inhibitor