Learn More
Cayman Chemical Resolvin D1 ELISA Standard, 100 ng
SDP
Catalog No. 502942479
Encompass_Preferred
Resolvin D1 ELISA Standard has been tested and formulated to work exclusively with Cayman’s Resolvin D1 ELISA Antiserum (Item No. 500380).
Catalog No. 50-294-2479 Supplier Cayman Chemical Supplier No. 400384100 NG
Resolvin D1 ELISA Standard has been tested and formulated to work exclusively with Cayman’s Resolvin D1 ELISA Antiserum (Item No. 500380).