Learn More
Cayman Chemical Rsperdon 10mg
SDP
Catalog No. 502953590
Encompass_Preferred
An atypical antipsychotic; binds to dopamine D2 receptors (Ki = 3 nM) and the serotonin (5-HT) receptor subtype 5-HT2A (Ki = 0.12 nM); also binds to dopamine D4, α1- and α2-adrenergic, 5-HT1C, 5-HT1D, and histamine H1 receptors (Kis = 7, 0.81, 7.3, 47, 52, and 2.1 nM, respectively); attenuates deficits in prepulse inhibition of the acoustic startle response, but not deficits in social interaction, in a rat neonatal ventral hippocampal lesion model of schizophrenia at 0.1 mg/kg per day
Catalog No. 50-295-3590 Supplier Cayman Chemical Supplier No. 1362910 MG
An atypical antipsychotic; binds to dopamine D2 receptors (Ki = 3 nM) and the serotonin (5-HT) receptor subtype 5-HT2A (Ki = 0.12 nM); also binds to dopamine D4, α1- and α2-adrenergic, 5-HT1C, 5-HT1D, and histamine H1 receptors (Kis = 7, 0.81, 7.3, 47, 52, and 2.1 nM, respectively); attenuates deficits in prepulse inhibition of the acoustic startle response, but not deficits in social interaction, in a rat neonatal ventral hippocampal lesion model of schizophrenia at 0.1 mg/kg per day