Learn More
Medchemexpress LLC RSPO1/R-spondin-1 H 10ug
SDP
Catalog No. 5000181710
Encompass_Preferred
RSPO1/R-spondin-1 is a secreted glycoprotein RSPO1/R-spondin-1 binds to LGR4/5/6 receptors and ZNRF3/E3 ubiquitin ligases inhibits Wnt receptor degradation stabilizes -catenin and promotes its nuclear translocation and activates target gene transcription RSPO1/R-spondin-1 Human promotes intestinal organoid survival induces pancreatic -cell proliferation to improve diabetes enhances osteogenic differentiation of mesenchymal stem cells to treat osteoporosis and inhibits bone erosion in arthritis models RSPO1/R-spondin-1 Protein Human (HEK293 His) is a recombinant RSPO1/R-spondin-1 protein expressed by HEK293 with a C-His tag
Catalog No. 50-001-81710 Supplier Medchemexpress LLC Supplier No. HYP7278410UG
RSPO1/R-spondin-1 is a secreted glycoprotein RSPO1/R-spondin-1 binds to LGR4/5/6 receptors and ZNRF3/E3 ubiquitin ligases inhibits Wnt receptor degradation stabilizes -catenin and promotes its nuclear translocation and activates target gene transcription RSPO1/R-spondin-1 Human promotes intestinal organoid survival induces pancreatic -cell proliferation to improve diabetes enhances osteogenic differentiation of mesenchymal stem cells to treat osteoporosis and inhibits bone erosion in arthritis models RSPO1/R-spondin-1 Protein Human (HEK293 His) is a recombinant RSPO1/R-spondin-1 protein expressed by HEK293 with a C-His tag