Learn More
Medchemexpress LLC RSPO1/R-spondin-1 H 50ug
SDP
Catalog No. 5000181806
Encompass_Preferred
RSPO1 or R-spondin-1 activates the canonical Wnt pathway through ligand binding to the LGR4-6 receptor forming a complex with phosphorylated LRP6 and Frizzled receptors This interaction upregulates target gene expression RSPO1/R-spondin-1 Protein Human (125a a HEK293 His) is the recombinant human-derived RSPO1/R-spondin-1 protein expressed by HEK293 with C-8 His labeled tag
Catalog No. 50-001-81806 Supplier Medchemexpress LLC Supplier No. HYP72784A50UG
RSPO1 or R-spondin-1 activates the canonical Wnt pathway through ligand binding to the LGR4-6 receptor forming a complex with phosphorylated LRP6 and Frizzled receptors This interaction upregulates target gene expression RSPO1/R-spondin-1 Protein Human (125a a HEK293 His) is the recombinant human-derived RSPO1/R-spondin-1 protein expressed by HEK293 with C-8 His labeled tag