Learn More
Medchemexpress LLC RSPO2/r-spondin-2 protein, mouse (HEK293, Fc) | >85.0% | 10 UG
SDP
Catalog No. 5000291961
Encompass_Preferred
Recombinant mouse R-spondin-2 (RSPO2) protein expressed in HEK293 cells and fused to a C-terminal mouse Fc tag, supplied as a 10 μg vial. The construct covers residues N24-G205 and is intended for in vitro biochemical and cell-based assays to modulate canonical Wnt/LGR signaling. Product documentation includes a datasheet and certificate of analysis.
Catalog No. 50-002-91961 Supplier Medchemexpress LLC Supplier No. HYP7601310UG
Recombinant mouse R-spondin-2 (RSPO2) protein expressed in HEK293 cells and fused to a C-terminal mouse Fc tag, supplied as a 10 μg vial. The construct covers residues N24-G205 and is intended for in vitro biochemical and cell-based assays to modulate canonical Wnt/LGR signaling. Product documentation includes a datasheet and certificate of analysis.