Learn More
Medchemexpress LLC Rspo2/r-spondin-2 protein, mouse (HEK293, Fc) | 92.0% | 100 UG
SDP
Catalog No. 5000291167
Encompass_Preferred
Recombinant mouse R-spondin-2 Fc-tagged protein produced in HEK293 cells for research on Wnt/β-catenin signaling. The Fc fusion enhances stability and facilitates detection or affinity purification. Supplied as a filtered PBS solution, formulated for storage at low temperature.
Catalog No. 50-002-91167 Supplier Medchemexpress LLC Supplier No. HYP76013100UG
Recombinant mouse R-spondin-2 Fc-tagged protein produced in HEK293 cells for research on Wnt/β-catenin signaling. The Fc fusion enhances stability and facilitates detection or affinity purification. Supplied as a filtered PBS solution, formulated for storage at low temperature.