Learn More
ACROBiosystems Recombinant Protein;Human R-Spondin 1 / RSPO1 (21-146) Protein;HEK293;50UG;RS6,RSPO1,RSPO-1,CRISTIN3,CRISTIN-3,FLJ40906,RP11-566C13.1,RSPO,R-spondin-1,Rspondin-1,Rspondin1,Rspondin
Catalog No. 501657720
Encompass_Preferred
Recombinant Protein;Human R-Spondin 1 / RSPO1 (21-146) Protein;50UG;Human R-Spondin 1, His Tag (RS6-H4220) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 146 (Accession # AAI14967).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Catalog No. 50-165-7720 Supplier ACROBiosystems Supplier No. RS6H422050UG
Recombinant Protein;Human R-Spondin 1 / RSPO1 (21-146) Protein;50UG;Human R-Spondin 1, His Tag (RS6-H4220) is expressed from human 293 cells (HEK293). It contains AA Ser 21 - Ala 146 (Accession # AAI14967).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 16.7 kDa. The protein migrates as 20-25 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.