Learn More
ACROBiosystems Recombinant Protein;Human R-Spondin 3 / RSPO3 (22-146) Protein;HEK293;1MG;RS6,RSPO3,RSPO-3,R-spondin 3,Rspondin 3,R-spondin-3,CRISTIN1,PWTSR,THSD2
Catalog No. 501657875
Encompass_Preferred
Recombinant Protein;Human R-Spondin 3 / RSPO3 (22-146) Protein;1MG;Human R-Spondin 3, His Tag (RS6-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 146 (Accession # AAH22367).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.3 kDa. The protein migrates as 18 kDa and 24 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.
Catalog No. 50-165-7875 Supplier ACROBiosystems Supplier No. RS6H52221MG
Recombinant Protein;Human R-Spondin 3 / RSPO3 (22-146) Protein;1MG;Human R-Spondin 3, His Tag (RS6-H5222) is expressed from human 293 cells (HEK293). It contains AA Gln 22 - Val 146 (Accession # AAH22367).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.3 kDa. The protein migrates as 18 kDa and 24 kDa under reducing (R) condition (SDS-PAGE) due to different glycosylation.