Learn More
New England Biolabs, Inc. RsrII – 2500 units
SDP
Catalog No. 50812139
Encompass
A restriction endonuclease that recognizes the sequence CG^GWC_CG.
Catalog No. 50-812-139 Supplier New England Biolabs, Inc. Supplier No. R0501L
A restriction endonuclease that recognizes the sequence CG^GWC_CG.