Learn More
New England Biolabs, Inc. RsrII – 500 units
SDP
Catalog No. 50812140
Encompass
A restriction endonuclease that recognizes the sequence CG^GWC_CG.
Catalog No. 50-812-140 Supplier New England Biolabs, Inc. Supplier No. R0501S
A restriction endonuclease that recognizes the sequence CG^GWC_CG.