Learn More
Pyramex™ RSSH32 Series Winter Wear Pullover Sweatshirt
Catalog No. 19555343 Shop All Pyramex Safety Products Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-555-343 Medium
19-555-342 Large
19-555-348 X-Large
19-555-344 2X-Large
19-555-345 3X-Large
19-555-346 4X-Large
19-555-347 5X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 19-555-343 Supplier Pyramex™ Supplier No. RSSH3210M
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
RSSH32 Series Winter Wear Pullover Sweatshirt is part of the Pyramex™ Safety Hi -Vis Winter Wear line.
  • 100% polyester 280 gsm fleece
  • Elastic knit at wrists and waist
  • 2 in. silver reflective material
  • Pull over type sweat shirt with hood
  • Mesh lined hood with adjustable pull cord
  • 1 large front pocket
Specifications
Size Medium
Gender Unisex
Material Polyester Fleece
Color Hi-vis Lime
Neck Style Hooded
Seam Type Sewn
Sleeve Style Long Sleeve
Cuff Style Elastic Knit
Disposable Reusable
No. of Pockets 1
Certifications/Compliance ANSI/ISEA 107-2015; Type R - Class 3
Tape Type China Brand Flat Silver
Thickness 280 gsm