Learn More
Cayman Chemical RSV F ProteIn ChImRIc Moncl
SDP
Catalog No. 502915420
Encompass_Preferred
Immunogen: Recombinant human-targeted RSV F • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) RSV • Cross Reactivity: (+) F protein • Applications: ELISA
Catalog No. 50-291-5420 Supplier Cayman Chemical Supplier No. 37173200 UG
Immunogen: Recombinant human-targeted RSV F • Host: Chimeric Monoclonal Antibody • Species Reactivity: (+) RSV • Cross Reactivity: (+) F protein • Applications: ELISA