Learn More
Cayman Chemical RSV F ProteIn EXrAClular D
SDP
Catalog No. 502915032
Encompass_Preferred
Source: Recombinant C-terminal His-tagged RSV F protein extracellular domain expressed in insect cells • Amino acids: 22-529 • MW: 57.8 kDa
Catalog No. 50-291-5032 Supplier Cayman Chemical Supplier No. 37020100 UG
Source: Recombinant C-terminal His-tagged RSV F protein extracellular domain expressed in insect cells • Amino acids: 22-529 • MW: 57.8 kDa