Learn More
Drg International Inc Rsv Igg Elisa
Supplier:  Drg International Inc EIA3507
Encompass_Preferred
An enzyme immunoassay for the qualitative and semiquantitative determination of IgG-class antibodies to Respiratory Syncytial Virus (RSV) in serum. Respiratory syncytial virus (RSV) is a negative-sense, enveloped RNA virus. It is the causative pathogen for the most common infection of the respiratory tract. Respiratory syncytial virus (RSV) infections usually occur during annual community outbreaks during the late fall, winter, or early spring months. The DRG RSV IgG ELISA Kit is a solid phase enzyme-linked immunosorbent assay (ELISA) Microtiter wells as a solid phase are coated with RSV antigen. Diluted patient specimens and ready-for-use controls are pipetted into these wells. During incubation RSV-specific antibodies of positive specimens and controls are bound to the immobilized antigens.
Catalog No. 50-194-7584