Learn More
Andwin Scientific RSV REAG 25ML 1CHL STRL,200/CS
SDP
Catalog No. NC0967004
Encompass_Preferred
The Reagent Reservoir features a volume capacity of 25 mL and a V bottom design that permits maximum recovery of reagents. This product has graduated pouring spots at all four corners and a wide base for heightened support. It is sold in a case of 200 sterile reservoirs.
Catalog No. NC0967004 Supplier Andwin Scientific Supplier No. S255200
The Reagent Reservoir features a volume capacity of 25 mL and a V bottom design that permits maximum recovery of reagents. This product has graduated pouring spots at all four corners and a wide base for heightened support. It is sold in a case of 200 sterile reservoirs.