Learn More
Enzo Life Sciences RSV Recombinant monoclonal antibody (RSHZ19) (Human IgG1 kappa) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS6550200
Encompass_Preferred
Clone: RSHZ19. Isotype: IgG1κ. Immunogen: This antibody was prepared by the humanization of an F protein-specific murine MAb, RSV19. More specifically, molecular techniques were used to insert the complementarity determining regions from murine RSV19 into a human IgG1 heavy and k light chain-variable domain framework. Species reactivity: Virus. Specificity: Recognizes the conserved fusion (F) protein of RSV. Applications: ELISA. Purity: Protein A affinity purified. Formulation: Liquid. In PBS containing 0.02% Proclin 300.
Catalog No. 50-201-3179