Learn More
Sino Biological Human respiratory syncytial virus (RSV) (A2) Fusion glycoprotein / RSV-F Antibody (HRP), Rabbit MAb
Supplier:  Sino Biological 11049R302H20
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human respiratory syncytial virus (RSV) (A2) Fusion glycoprotein / RSV-F ( Catalog#11049-V08B; P03420.1; Met1-Thr529), and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-240-8208