Learn More
Sino Biological Human respiratory syncytial virus (RSV) (A2) Fusion glycoprotein / RSV-F Antibody, Rabbit MAb
Supplier:  Sino Biological 11049R30220
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant Human respiratory syncytial virus (RSV) (A2) Fusion glycoprotein / RSV-F ( Catalog#11049-V08B; P03420.1; Met1-Thr529).
Catalog No. 50-237-1174