Learn More
BTNX Inc RSV Test Cassette - 20pk
Catalog No. NC3361245
Encompass_Preferred
The Rapid Response™ RSV Test Cassette is an in vitro diagnostic test for the qualitative detection of Respiratory Syncytial Virus antigens (viral fusion protein) in nasopharyngeal swab, nasopharyngeal aspirate or nasopharyngeal wash specimens using the rapid immuno-chromatographic method. It is intended to aid in the rapid differential diagnosis of Respiratory Syncytial Virus viral infections. 
This item is not returnable. View return policy
Catalog No. NC3361245 Supplier BTNX Inc Supplier No. RSV19C20
The Rapid Response™ RSV Test Cassette is an in vitro diagnostic test for the qualitative detection of Respiratory Syncytial Virus antigens (viral fusion protein) in nasopharyngeal swab, nasopharyngeal aspirate or nasopharyngeal wash specimens using the rapid immuno-chromatographic method. It is intended to aid in the rapid differential diagnosis of Respiratory Syncytial Virus viral infections. 