Learn More
TARGETMOL CHEMICALS INC RSV604 25MG
SDP
Catalog No. 502579206
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RSV604 ((S)-1-(2-Fluorophenyl)-3-(2-oxo-5-phenyl-23-dihydro-1H-benzo[e][14]diazepin-3-yl)urea) is an inhibitor of respiratory syncytial virus (RSV) that binds to RSV nucleoprotein (Kd = 1.31 uM)purity: 98%
Catalog No. 50-257-9206 Supplier TARGETMOL CHEMICALS INC Supplier No. T735325MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. RSV604 ((S)-1-(2-Fluorophenyl)-3-(2-oxo-5-phenyl-23-dihydro-1H-benzo[e][14]diazepin-3-yl)urea) is an inhibitor of respiratory syncytial virus (RSV) that binds to RSV nucleoprotein (Kd = 1.31 uM)purity: 98%