Learn More
Medchemexpress LLC HY-12993A 5mg Medchemexpress, RSV604 (racemate) CAS:676128-62-4 Purity:>98%
SDP
Catalog No. 501932645
Encompass_Preferred
Medchemexpress, HY-12993A 5mg RSV604 (racemate) CAS:676128-62-4 RSV604 racemate is a racemic mixture, shows less potency against strains of respiratory syncytial virus (RSV) than the S-isomer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2645 Supplier Medchemexpress LLC Supplier No. HY12993A5MG
Medchemexpress, HY-12993A 5mg RSV604 (racemate) CAS:676128-62-4 RSV604 racemate is a racemic mixture, shows less potency against strains of respiratory syncytial virus (RSV) than the S-isomer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.