Learn More
Medchemexpress LLC HY-103238 5mg Medchemexpress, RSVA405 CAS:140405-36-3 Purity:>98%
SDP
Catalog No. 502080660
Encompass_Preferred
Medchemexpress, HY-103238 5mg RSVA405 CAS:140405-36-3 RSVA405 is a potent, orally active activator of AMPK, with an EC50 of 1 μM. RSVA405 facilitates CaMKKβ-dependent activation of AMPK, inhibits mTOR, and promotes autophagy to increase Aβ degradation. RSVA405 has anti-inflammatory effects through the inhibition of STAT3 function. RSVA405 also can be used for the research of obesity[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0660 Supplier Medchemexpress LLC Supplier No. HY1032385MG
Medchemexpress, HY-103238 5mg RSVA405 CAS:140405-36-3 RSVA405 is a potent, orally active activator of AMPK, with an EC50 of 1 μM. RSVA405 facilitates CaMKKβ-dependent activation of AMPK, inhibits mTOR, and promotes autophagy to increase Aβ degradation. RSVA405 has anti-inflammatory effects through the inhibition of STAT3 function. RSVA405 also can be used for the research of obesity[4]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.