Learn More
Cole-Parmer RT 5 Position Stirring Hot Plate, 115V
Catalog No. NC1953787 Shop All Cole-Parmer Products
Encompass
Wear-free/maintenance-free magnetic coils for consistent and silent operation. Stir consistently in all positions. Adjust speed in 10-rpm increments-speed remains constant.
Catalog No. NC1953787 Supplier Cole-Parmer Supplier No. 0467194
Wear-free/maintenance-free magnetic coils for consistent and silent operation. Stir consistently in all positions. Adjust speed in 10-rpm increments-speed remains constant.