Learn More
Biocare Medical, LLC RT DETECTION ANTI-MS-CD31 6ML
SDP
Supplier:  Biocare Medical, LLC RT517SK
Encompass_Preferred
The Rat Detection Kit for Anti-Mouse CD31 is for detection of a rat anti-mouse CD31 primary antibody (sold seperately) on mouse tissues. It contains blockers, digestion enzyme, antibody diluent, HRP-Polymer, DAB chromogen and hematoxylin.
Catalog No. 50-832-86