Learn More
AdipoGen Rat IgG1 Isotype Control (preservative free) (50 µg)
SDP
Supplier:  AdipoGen AG35B0012PFC050
Encompass_Preferred
Monoclonal antibody. Applications: FACS, FUNC, ICC, IHC, IP. Clone: KLH/G1-2-2. Isotype: Rat IgG1kappa. Liquid. In PBS. Purity: >95% (SDS-PAGE). Negative control in assays where preservative free rat IgG1 antibodies are used.
Catalog No. 50-205-3058