Learn More
PREDICT HEALTH HBV viral load (Quantitative) RT-PCR kit is a quantitative RT PCR based diagnostic kit for detection of HBV DNA in human plasma/serum or blood samples. 100 reactions
Supplier:  PREDICT HEALTH DVZ 30091
Encompass_Preferred
HBV viral load (Quantitative) RT-PCR kit is an in vitro nucleic acid amplification test for the detection & quantitation of the HBV DNA in human plasma/serum or blood samples. This test kit utilizes the real time RT-PCR after the extraction of pure nucleic acid from the plasma/serum samples. The viral load is measured using a range of four quantification calibrators provided in the kit
Catalog No. NC2891005