Learn More
USA Scientific Inc TempPlate RT Select optically clear film, inset cut
Catalog No. NC9844216
Encompass
TempPlate RT Select optically clear real-time PCR film with strong silicone adhesive, -70C to +100C, non-sterile, 100/box
Catalog No. NC9844216 Supplier USA Scientific Inc Supplier No. 29217800
TempPlate RT Select optically clear real-time PCR film with strong silicone adhesive, -70C to +100C, non-sterile, 100/box