Learn More
Apexbio Technology LLC RT SuperMix for qPCR, 100 rxn (20 uL/rxn). Cas: MFCD: NA. High efficiency reverse transcription reaction premixed solution for two-step RT-qPCR method
SDP
Catalog No. 501998232
Encompass_Preferred
RT SuperMix for qPCR is the premixed solution for reverse transcription reaction, based on Reverse Transcriptase (Cat. No. K1071). Reverse Transcriptase is a new enzyme obtained through genetic engineering based on M-MLV (RNase H-) Reverse Transcriptase. In comparison, Reverse Transcriptase reduces RNase H activity and increases thermal stability. Reverse Transcriptase can withstand higher reaction temperatures and is suitable for reverse transcription of RNA templates with complex secondary structures.
Catalog No. 50-199-8232 Supplier Apexbio Technology LLC Supplier No. K1074100
RT SuperMix for qPCR is the premixed solution for reverse transcription reaction, based on Reverse Transcriptase (Cat. No. K1071). Reverse Transcriptase is a new enzyme obtained through genetic engineering based on M-MLV (RNase H-) Reverse Transcriptase. In comparison, Reverse Transcriptase reduces RNase H activity and increases thermal stability. Reverse Transcriptase can withstand higher reaction temperatures and is suitable for reverse transcription of RNA templates with complex secondary structures.