Learn More
ATCC RT4; Bladder Papilloma; Human (Homo sapiens)
Catalog No. 502383572
Encompass
RT4 is a cell line exhibiting epithelial morphology that was isolated from urinary bladder tissue derived from a 63-year-old, White, male patient with transitional cell papilloma. Use these cells in your immuno-oncology research.
Catalog No. 50-238-3572 Supplier ATCC Supplier No. HTB2
RT4 is a cell line exhibiting epithelial morphology that was isolated from urinary bladder tissue derived from a 63-year-old, White, male patient with transitional cell papilloma. Use these cells in your immuno-oncology research.